siRNA Details
| Gene Symbol: Gfra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012959.1 | NP_037091.1 | 5 | 988 | ||
| XM_008760512.1 | XP_008758734.1 | 6 | 1131 | |||
| XM_008760513.1 | XP_008758735.1 | 6 | 995 | |||
| XM_008760514.1 | XP_008758736.1 | 4 | 1011 | |||
| GenBank mRNA | AJ002072.1 | - | 4 | 717 | ||
| U59486.1 | - | 5 | 1033 | |||
| U97142.1 | - | 5 | 988 | |||

Gfra1
Genomic Map