siRNA Details
Gene Symbol: Man2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012979.2 | NP_037111.2 | 7 | 2111 | ||
GenBank mRNA | AY325162.1 | - | 126 | |||
M24353.1 | - | 7 | 1028 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Man2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012979.2 | NP_037111.2 | 7 | 2111 | ||
GenBank mRNA | AY325162.1 | - | 126 | |||
M24353.1 | - | 7 | 1028 |