siRNA Details
| Gene Symbol: Vps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105718.2 | NP_001099188.2 | 15 | 1933 | ||
| GenBank mRNA | BC169004.1 | - | 10 | 1374 | ||
| CK596387.1 | - | 4 | 498 | |||
| EV778075.1 | - | 2 | 253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps35| Gene Symbol: Vps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105718.2 | NP_001099188.2 | 15 | 1933 | ||
| GenBank mRNA | BC169004.1 | - | 10 | 1374 | ||
| CK596387.1 | - | 4 | 498 | |||
| EV778075.1 | - | 2 | 253 | |||