siRNA Details
| Gene Symbol: Nfia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012988.2 | NP_037120.1 | 5 | 1079 | ||
| GenBank mRNA | AB060652.1 | - | 987 | |||
| AF112455.1 | - | 1159 | ||||
| D78017.1 | - | 5 | 1079 | |||
| D78018.1 | - | 4 | 1008 | |||
| D78020.1 | - | 5 | 501 | |||
| X13167.1 | - | 5 | 921 | |||

Nfia
Genomic Map