siRNA Details
| Gene Symbol: Nid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213954.8 | XP_213954.4 | 13 | 2755 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nid1| Gene Symbol: Nid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213954.8 | XP_213954.4 | 13 | 2755 | ||