siRNA Details
| Gene Symbol: Nog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012990.1 | NP_037122.1 | 1 | 333 | ||
| GenBank mRNA | BF404195.1 | - | 1 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nog| Gene Symbol: Nog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012990.1 | NP_037122.1 | 1 | 333 | ||
| GenBank mRNA | BF404195.1 | - | 1 | 332 | ||