siRNA Details
| Gene Symbol: Ppia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017101.1 | NP_058797.1 | 5 | 440 | ||
| GenBank mRNA | BC059141.1 | - | 5 | 445 | 6919594 | |
| BC091153.1 | - | 5 | 416 | 7308693 | ||
| FQ210268.1 | - | 5 | 441 | |||
| FQ210843.1 | - | 5 | 443 | |||
| FQ222826.1 | - | 5 | 441 | |||
| FQ223330.1 | - | 5 | 442 | |||
| FQ223366.1 | - | 5 | 441 | |||
| FQ223500.1 | - | 5 | 441 | |||
| FQ227889.1 | - | 5 | 441 | |||
| FQ228411.1 | - | 5 | 441 | |||
| FQ228450.1 | - | 5 | 441 | |||
| FQ233894.1 | - | 5 | 440 | |||
| FQ234872.1 | - | 5 | 441 | |||
| M19533.1 | - | 5 | 440 | |||
| M25637.1 | - | 5 | 433 | |||
| Gene Symbol: LOC100360977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250801.2 | XP_006250863.1 | 1 | 437 | ||

Ppia
Genomic Map