siRNA Details
| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033679.1 | NP_001028851.2 | 1 | 137 | ||
| XM_008768942.1 | XP_008767164.1 | 1 | 230 | |||
| GenBank mRNA | BC104679.1 | - | 1 | 137 | 7456971 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkar1b| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033679.1 | NP_001028851.2 | 1 | 137 | ||
| XM_008768942.1 | XP_008767164.1 | 1 | 230 | |||
| GenBank mRNA | BC104679.1 | - | 1 | 137 | 7456971 | |