siRNA Details
| Gene Symbol: Rab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013017.1 | NP_037149.1 | 488 | |||
| GenBank mRNA | M83676.1 | - | 3, 4 | 283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab12| Gene Symbol: Rab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013017.1 | NP_037149.1 | 488 | |||
| GenBank mRNA | M83676.1 | - | 3, 4 | 283 | ||