siRNA Details
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013025.2 | NP_037157.2 | 1 | 93 | ||
GenBank mRNA | EF121994.1 | - | 1 | 24 | ||
EF121995.1 | - | 1 | 24 | |||
U22414.1 | - | 1 | 93 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013025.2 | NP_037157.2 | 1 | 93 | ||
GenBank mRNA | EF121994.1 | - | 1 | 24 | ||
EF121995.1 | - | 1 | 24 | |||
U22414.1 | - | 1 | 93 |