siRNA Details
| Gene Symbol: Sele | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138879.1 | NP_620234.1 | 4, 5 | 718 | ||
| XM_008769608.1 | XP_008767830.1 | 5, 6 | 1100 | |||
| GenBank mRNA | L25527.1 | - | 4, 5 | 718 | ||
| X69981.1 | - | 4, 5 | 769 | |||

Sele
Genomic Map