siRNA Details
| Gene Symbol: Tnfrsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013049.1 | NP_037181.1 | 5 | 481 | ||
| GenBank mRNA | X17037.1 | - | 5 | 481 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf4| Gene Symbol: Tnfrsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013049.1 | NP_037181.1 | 5 | 481 | ||
| GenBank mRNA | X17037.1 | - | 5 | 481 | ||