siRNA Details
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013062.1 | NP_037194.1 | 10 | 1639 | ||
GenBank mRNA | BC087029.1 | - | 10 | 1615 | 7124961 | |
U93306.1 | - | 10 | 1639 | |||
U93307.1 | - | 10 | 1639 |
