siRNA Details
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105724.1 | NP_001099194.1 | 7 | 675 | ||
GenBank mRNA | D30734.1 | - | 7 | 719 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105724.1 | NP_001099194.1 | 7 | 675 | ||
GenBank mRNA | D30734.1 | - | 7 | 719 |