siRNA Details
| Gene Symbol: Rasa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105724.1 | NP_001099194.1 | 8, 9 | 776 | ||
| GenBank mRNA | D30734.1 | - | 8, 9 | 820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasa2| Gene Symbol: Rasa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105724.1 | NP_001099194.1 | 8, 9 | 776 | ||
| GenBank mRNA | D30734.1 | - | 8, 9 | 820 | ||