siRNA Details
| Gene Symbol: Asns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013079.2 | NP_037211.2 | 7, 8 | 1207 | ||
| XM_006236089.2 | XP_006236151.1 | 7, 8 | 1200 | |||
| GenBank mRNA | BC081719.1 | - | 7, 8 | 1207 | 7132312 | |
| U07201.1 | - | 7, 8 | 1151 | |||
| U07202.1 | - | 7, 8 | 1151 | |||

Asns
Genomic Map