siRNA Details
| Gene Symbol: Ptprz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013080.2 | NP_037212.2 | 20 | 5689 | ||
| XM_006236137.2 | XP_006236199.1 | 19 | 5707 | |||
| XM_006236138.2 | XP_006236200.1 | 20 | 3169 | |||
| XM_006236139.2 | XP_006236201.1 | 19 | 3148 | |||
| GenBank mRNA | U09357.1 | - | 21 | 5689 | ||

Ptprz1
Genomic Map