siRNA Details
| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177593.1 | NP_001171064.1 | 12 | 1595 | ||
| NM_013088.2 | NP_037220.2 | 12 | 1583 | |||
| GenBank mRNA | D83016.1 | - | 6 | 632 | ||
| U05963.1 | - | 12 | 1526 | |||
| U09307.1 | - | 12 | 1508 | |||

Ptpn11
Genomic Map