siRNA Details
| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013090.2 | NP_037222.2 | 3 | 318 | ||
| GenBank mRNA | AF498262.1 | - | 2 | 184 | ||
| BC092206.1 | - | 3 | 216 | 7313343 | ||
| CB582258.1 | - | 3 | 318 | |||
| CO567426.1 | - | 347 | ||||
| EF653274.1 | - | 3 | 243 | |||
| EF653275.1 | - | 3 | 243 | |||
| M24104.1 | - | 3 | 281 | |||
| U74621.1 | - | 3 | 216 | |||

Vamp1
Genomic Map