siRNA Details
| Gene Symbol: Cma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013092.1 | NP_037224.1 | 2 | 202 | ||
| GenBank mRNA | D38495.1 | - | 2 | 202 | ||
| U67908.1 | - | 2 | 167 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cma1| Gene Symbol: Cma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013092.1 | NP_037224.1 | 2 | 202 | ||
| GenBank mRNA | D38495.1 | - | 2 | 202 | ||
| U67908.1 | - | 2 | 167 | |||