siRNA Details
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013092.1 | NP_037224.1 | 5 | 764 | ||
GenBank mRNA | D38495.1 | - | 5 | 764 | ||
U67908.1 | - | 5 | 729 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013092.1 | NP_037224.1 | 5 | 764 | ||
GenBank mRNA | D38495.1 | - | 5 | 764 | ||
U67908.1 | - | 5 | 729 |