siRNA Details
| Gene Symbol: Nkx2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013093.1 | NP_037225.1 | 2 | 1073 | ||
| XM_006240116.2 | XP_006240178.1 | 3 | 1354 | |||
| XM_006240117.2 | XP_006240179.1 | 3 | 1427 | |||
| XM_006240118.2 | XP_006240180.1 | 3 | 1938 | |||
| GenBank mRNA | AB894119.1 | - | 2 | 1072 | ||
| X53858.1 | - | 2 | 1073 | |||

Nkx2-1
Genomic Map