siRNA Details
| Gene Symbol: Plin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013094.1 | NP_037226.1 | 4 | 562 | ||
| XM_008759499.1 | XP_008757721.1 | 5 | 730 | |||
| GenBank mRNA | L26043.1 | - | 4 | 562 | ||
| L26044.1 | - | 4 | 562 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plin1| Gene Symbol: Plin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013094.1 | NP_037226.1 | 4 | 562 | ||
| XM_008759499.1 | XP_008757721.1 | 5 | 730 | |||
| GenBank mRNA | L26043.1 | - | 4 | 562 | ||
| L26044.1 | - | 4 | 562 | |||