siRNA Details
| Gene Symbol: Otx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013109.2 | NP_037241.1 | 3 | 701 | ||
| GenBank mRNA | L32602.1 | - | 3 | 702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otx1| Gene Symbol: Otx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013109.2 | NP_037241.1 | 3 | 701 | ||
| GenBank mRNA | L32602.1 | - | 3 | 702 | ||