siRNA Details
| Gene Symbol: Selp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013114.1 | NP_037246.1 | 4 | 739 | ||
| XM_006250150.2 | XP_006250212.1 | 5 | 853 | |||
| GenBank mRNA | BC101919.1 | - | 4 | 729 | 7935390 | |
| L23088.1 | - | 4 | 739 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selp| Gene Symbol: Selp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013114.1 | NP_037246.1 | 4 | 739 | ||
| XM_006250150.2 | XP_006250212.1 | 5 | 853 | |||
| GenBank mRNA | BC101919.1 | - | 4 | 729 | 7935390 | |
| L23088.1 | - | 4 | 739 | |||