siRNA Details
Gene Symbol: Guca2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013118.1 | NP_037250.1 | 2, 3 | 292 | ||
GenBank mRNA | M93005.1 | - | 2, 3 | 292 | ||
M95493.1 | - | 2, 3 | 292 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Guca2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013118.1 | NP_037250.1 | 2, 3 | 292 | ||
GenBank mRNA | M93005.1 | - | 2, 3 | 292 | ||
M95493.1 | - | 2, 3 | 292 |