siRNA Details
| Gene Symbol: Igfbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013122.2 | NP_037254.2 | 4 | 941 | ||
| GenBank mRNA | BC092570.1 | - | 4 | 941 | 7308779 | |
| J04486.1 | - | 4 | 1133 | |||
| M31672.1 | - | 4 | 969 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp2| Gene Symbol: Igfbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013122.2 | NP_037254.2 | 4 | 941 | ||
| GenBank mRNA | BC092570.1 | - | 4 | 941 | 7308779 | |
| J04486.1 | - | 4 | 1133 | |||
| M31672.1 | - | 4 | 969 | |||