siRNA Details
| Gene Symbol: Cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013128.1 | NP_037260.1 | 5 | 1004 | ||
| GenBank mRNA | BC085762.1 | - | 5 | 1003 | 7193151 | |
| J04625.1 | - | 5 | 959 | |||
| M31602.1 | - | 5 | 975 | |||
| X51406.1 | - | 5 | 1004 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpe| Gene Symbol: Cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013128.1 | NP_037260.1 | 5 | 1004 | ||
| GenBank mRNA | BC085762.1 | - | 5 | 1003 | 7193151 | |
| J04625.1 | - | 5 | 959 | |||
| M31602.1 | - | 5 | 975 | |||
| X51406.1 | - | 5 | 1004 | |||