siRNA Details
| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013129.2 | NP_037261.1 | 4, 5 | 371 | ||
| XM_008772363.1 | XP_008770585.1 | 4, 5 | 325 | |||
| GenBank mRNA | AF015718.1 | - | 4, 5 | 323 | ||
| AF015719.1 | - | 4, 5 | 371 | |||
| BC097340.1 | - | 5, 6 | 1175 | |||
| U69272.1 | - | 916 | ||||

Il15
Genomic Map