siRNA Details
Gene Symbol: Clps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013139.1 | NP_037271.1 | 1 | 72 | ||
GenBank mRNA | M33333.1 | - | 1 | 31 | ||
M58370.1 | - | 1 | 72 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013139.1 | NP_037271.1 | 1 | 72 | ||
GenBank mRNA | M33333.1 | - | 1 | 31 | ||
M58370.1 | - | 1 | 72 |