siRNA Details
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013141.2 | NP_037273.2 | 7 | 1044 | ||
XM_006256166.2 | XP_006256228.1 | 7 | 1856 | |||
XM_006256167.2 | XP_006256229.1 | 6 | 1963 | |||
XM_006256168.2 | XP_006256230.1 | 6 | 2858 | |||
XM_008772797.1 | XP_008771019.1 | 6 | 2091 | |||
GenBank mRNA | AJ306400.1 | - | 5 | 810 | ||
U40064.1 | - | 7 | 1065 | |||
U75918.1 | - | 7 | 1058 |
