siRNA Details
| Gene Symbol: Col12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766422.1 | XP_008764644.1 | 8 | 4620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col12a1| Gene Symbol: Col12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766422.1 | XP_008764644.1 | 8 | 4620 | ||