siRNA Details
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013167.2 | NP_037299.1 | 7 | 1022 | ||
| GenBank mRNA | AB006614.1 | - | 6 | 894 | ||
| AF030163.1 | - | 6 | 909 | |||
| AF035943.1 | - | 6 | 905 | |||
| BC072546.1 | - | 7 | 1022 | 7105743 | ||
| U92069.1 | - | 7 | 1029 | |||

Ucp3
Genomic Map