siRNA Details
| Gene Symbol: Tcf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013176.2 | NP_037308.1 | 18 | 1882 | ||
| XM_006243345.1 | XP_006243407.1 | 18 | 1737 | |||
| XM_006243346.1 | XP_006243408.1 | 18 | 1737 | |||
| XM_006243347.1 | XP_006243409.1 | 17 | 1668 | |||
| XM_006243348.1 | XP_006243410.1 | 17 | 1665 | |||
| XM_006243349.1 | XP_006243411.1 | 17 | 1665 | |||
| XM_006243350.1 | XP_006243412.1 | 17 | 1662 | |||
| XM_006243351.1 | XP_006243413.1 | 16 | 1593 | |||
| XM_006243352.2 | XP_006243414.1 | 11 | 1531 | |||
| XM_006243353.2 | XP_006243415.1 | 11 | 1528 | |||
| XM_006243354.2 | XP_006243416.1 | 10 | 1456 | |||
| XM_008766218.1 | XP_008764440.1 | 16 | 5421 | |||
| XM_008766219.1 | XP_008764441.1 | 10 | 1459 | |||
| GenBank mRNA | AJ973226.1 | - | 11 | 1256 | ||
| L09656.1 | - | 18 | 1882 | |||

Tcf12
Genomic Map