siRNA Details
| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013192.2 | NP_037324.1 | 2 | 721 | ||
| XM_008768567.1 | XP_008766789.1 | 1 | 507 | |||
| GenBank mRNA | AB073753.1 | - | 2 | 721 | ||
| AB073754.1 | - | 2 | 721 | |||
| AB073755.1 | - | 2 | 721 | |||
| AB073756.1 | - | 2 | 721 | |||
| AY095171.1 | - | 1 | 510 | |||
| U21087.1 | - | 2 | 721 | |||

Kcnj6
Genomic Map