siRNA Details
Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147139.2 | NP_671480.2 | 3 | 358 | ||
GenBank mRNA | CB731093.1 | - | 3 | 328 | ||
EV772196.1 | - | 2 | 211 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147139.2 | NP_671480.2 | 3 | 358 | ||
GenBank mRNA | CB731093.1 | - | 3 | 328 | ||
EV772196.1 | - | 2 | 211 |