siRNA Details
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271054.1 | NP_001257983.1 | 12, 13 | 1734 | ||
| XM_006240151.2 | XP_006240213.1 | 12, 13 | 1784 | |||
| XM_006240152.2 | XP_006240214.1 | 13, 14 | 1864 | |||
| XM_006240153.2 | XP_006240215.1 | 12, 13 | 2208 | |||

Frmd6
Genomic Map