siRNA Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147211.1 | NP_671744.1 | 7 | 1490 | ||
| XM_008762874.1 | XP_008761096.1 | 8 | 2027 | |||
| XM_008762875.1 | XP_008761097.1 | 8 | 4354 | |||
| XM_008762876.1 | XP_008761098.1 | 8 | 2027 | |||
| XM_008762877.1 | XP_008761099.1 | 8 | 2025 | |||
| GenBank mRNA | U25281.1 | - | 7 | 1490 | ||
| U31159.1 | - | 8 | 1592 | |||

Wipf3
Genomic Map