siRNA Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147211.1 | NP_671744.1 | 7, 8 | 1521 | ||
| XM_008762877.1 | XP_008761099.1 | 8, 9 | 2056 | |||
| GenBank mRNA | U25281.1 | - | 7, 8 | 1521 | ||
| U31159.1 | - | 8, 9 | 1623 | |||

Wipf3
Genomic Map