siRNA Details
| Gene Symbol: Zfp335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762559.1 | XP_008760781.1 | 13 | 2576 | ||
| XM_008762560.1 | XP_008760782.1 | 14 | 3250 | |||
| XM_008762561.1 | XP_008760783.1 | 13 | 2573 | |||
| XM_008762562.1 | XP_008760784.1 | 13 | 2576 | |||
| XM_008762563.1 | XP_008760785.1 | 13 | 2576 | |||
| XM_008762564.1 | XP_008760786.1 | 13 | 2573 | |||
| XM_008762565.1 | XP_008760787.1 | 13 | 2576 | |||
| GenBank mRNA | AY079168.1 | - | 5 | 348 | ||

Zfp335
Genomic Map