siRNA Details
| Gene Symbol: Gpr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152242.2 | NP_689448.2 | 13 | 1973 | ||
| XM_006255066.2 | XP_006255128.1 | 13 | 3025 | |||
| XM_008772264.1 | XP_008770486.1 | 13 | 2175 | |||
| XM_008772265.1 | XP_008770487.1 | 14 | 2439 | |||
| XM_008772266.1 | XP_008770488.1 | 13 | 3685 | |||
| XM_008772267.1 | XP_008770489.1 | 13 | 3948 | |||
| XM_008772268.1 | XP_008770490.1 | 14 | 2272 | |||
| XM_008772269.1 | XP_008770491.1 | 13 | 3054 | |||
| XM_008772270.1 | XP_008770492.1 | 13 | 3530 | |||
| GenBank mRNA | AF529886.1 | - | 13 | 1974 | ||
| CK482497.1 | - | 3 | 169 | |||
More Information
Set Membership: |
|

Gpr56
Genomic Map