siRNA Details
Gene Symbol: Gpr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152242.2 | NP_689448.2 | 4, 5 | 903 | ||
XM_006255066.2 | XP_006255128.1 | 4, 5 | 1955 | |||
XM_008772264.1 | XP_008770486.1 | 4, 5 | 1105 | |||
XM_008772265.1 | XP_008770487.1 | 5, 6 | 1369 | |||
XM_008772266.1 | XP_008770488.1 | 4, 5 | 2615 | |||
XM_008772267.1 | XP_008770489.1 | 4, 5 | 2878 | |||
XM_008772268.1 | XP_008770490.1 | 5, 6 | 1202 | |||
XM_008772269.1 | XP_008770491.1 | 4, 5 | 1984 | |||
XM_008772270.1 | XP_008770492.1 | 4, 5 | 2460 | |||
GenBank mRNA | AF529886.1 | - | 4, 5 | 904 | ||
CV116515.1 | - | 2, 3 | 359 | |||
EX488169.1 | - | 1, 2 | 14 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
