siRNA Details
| Gene Symbol: Csdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170542.1 | NP_001164013.1 | 2 | 332 | ||
| GenBank mRNA | CO405729.1 | - | 2 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csdc2| Gene Symbol: Csdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170542.1 | NP_001164013.1 | 2 | 332 | ||
| GenBank mRNA | CO405729.1 | - | 2 | 332 | ||