siRNA Details
| Gene Symbol: Csdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170542.1 | NP_001164013.1 | 4 | 731 | ||
| GenBank mRNA | DY318921.1 | - | 2 | 131 | ||
| X89962.1 | - | 4 | 630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csdc2| Gene Symbol: Csdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170542.1 | NP_001164013.1 | 4 | 731 | ||
| GenBank mRNA | DY318921.1 | - | 2 | 131 | ||
| X89962.1 | - | 4 | 630 | |||