siRNA Details
| Gene Symbol: Fadd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152937.2 | NP_690920.1 | 2 | 475 | ||
| GenBank mRNA | AF406779.1 | - | 2 | 475 | ||
| AJ441127.1 | - | 500 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fadd| Gene Symbol: Fadd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152937.2 | NP_690920.1 | 2 | 475 | ||
| GenBank mRNA | AF406779.1 | - | 2 | 475 | ||
| AJ441127.1 | - | 500 | ||||