siRNA Details
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153309.2 | NP_695221.1 | 3 | 515 | ||
XM_008768680.1 | XP_008766902.1 | 3 | 580 | |||
GenBank mRNA | AB083195.1 | - | 2 | 490 | ||
BC087650.1 | - | 3 | 515 | 7310399 |
Gene Symbol: LOC689439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761010.1 | XP_008759232.1 | 1 | 558 |
