siRNA Details
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153309.2 | NP_695221.1 | 3 | 568 | ||
XM_008768680.1 | XP_008766902.1 | 3 | 633 | |||
GenBank mRNA | AB083195.1 | - | 2 | 543 | ||
BC087650.1 | - | 3 | 568 | 7310399 |
