siRNA Details
| Gene Symbol: Mina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153309.2 | NP_695221.1 | 5 | 767 | ||
| XM_008768680.1 | XP_008766902.1 | 5 | 832 | |||
| GenBank mRNA | AB083195.1 | - | 4 | 742 | ||
| BC087650.1 | - | 5 | 767 | 7310399 | ||

Mina
Genomic Map