siRNA Details
| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153315.2 | NP_695227.2 | 7, 8 | 847 | ||
| GenBank mRNA | AB062758.1 | - | 7, 8 | 663 | ||
| BC070961.1 | - | 7, 8 | 847 | 7099044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs4| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153315.2 | NP_695227.2 | 7, 8 | 847 | ||
| GenBank mRNA | AB062758.1 | - | 7, 8 | 663 | ||
| BC070961.1 | - | 7, 8 | 847 | 7099044 | ||