siRNA Details
| Gene Symbol: Slc35e4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153316.2 | NP_695228.2 | 2 | 1380 | ||
| GenBank mRNA | AF182714.1 | - | 3 | 1103 | ||
| BC085693.1 | - | 2 | 1380 | 7191218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35e4| Gene Symbol: Slc35e4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153316.2 | NP_695228.2 | 2 | 1380 | ||
| GenBank mRNA | AF182714.1 | - | 3 | 1103 | ||
| BC085693.1 | - | 2 | 1380 | 7191218 | ||